missing translation for 'onlineSavingsMsg'
Learn More

N,N,N-Trimethylglycine N-Hydroxysuccinimide Ester Bromide, TRC

Product Code. 30450838
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450838 10 mg 10mg
30449870 50 mg -
30449559 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450838 Supplier Toronto Research Chemicals Supplier No. TRCT79605010MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42014-55-1
Molecular Formula C9 H15 N2 O4 . Br
Molecular Weight (g/mol) 295.13
IUPAC Name [2-(2,5-dioxopyrrolidin-1-yl)oxy-2-oxoethyl]-trimethylazanium;bromide
SMILES [Br-].C[N+](C)(C)CC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H15N2O4.BrH/c1-11(2,3)6-9(14)15-10-7(12)4-5-8(10)13;/h4-6H2,1-3H3;1H/q+1;/p-1
Formula Weight 294.0215
Chemical Name or Material N,N,N-Trimethylglycine-N-Hydroxysuccinimide Ester, Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.