missing translation for 'onlineSavingsMsg'
Learn More

N,N,N-Trimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanaminium Chloride, Mikromol™

Product Code. 30049278
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049278 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049278 Supplier Mikromol Supplier No. MM1313.130025
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N,N,N-Trimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanaminium Chloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N,N-Trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanaminium Chloride
Analyte or Component Names N,N,N-Trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanaminium Chloride
Shipping Condition Room Temperature
Molecular Formula C16 H22 N5 . Cl
Certifications/Compliance ISO 9001
Synonym N,N,N-Trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanaminium Chloride, 2-{5-[(1H-1,2,4-Triazol-1-yl)methyl]-1H-indol-3-yl}-N,N,N-trimethylethan-1-aminium Chloride, Trimethyl-[2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethyl]ammonium Chloride
InChI Formula InChI=1S/C16H22N5.ClH/c1-21(2,3)7-6-14-9-18-16-5-4-13(8-15(14)16)10-20-12-17-11-19-20;/h4-5,8-9,11-12,18H,6-7,10H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].C[N+](C)(C)CCc1c[nH]c2ccc(Cn3cncn3)cc12
IUPAC Name trimethyl-[2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethyl]azanium;chloride
Molecular Weight (g/mol) 319.83
Quantity 25 mg
Formula Weight 319.1564 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.