missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis

Product Code. 10398863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398863 2.5 kg Plastic Bottle 2.5kg
10760102 5 g Glass bottle 5g
10168180 50 g Glass bottle 50g
10137560 100 g Glass bottle 100g
10005660 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10398863 Supplier Thermo Scientific Chemicals Supplier No. 420580025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-18-9 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -55.0°C
Color Colorless to Yellow
Density 0.7700g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Bottle
Linear Formula (CH3)2NCH2CH2N(CH3)2
Refractive Index 1.4169 to 1.4189
Quantity 2.5 kg
Beilstein 04, 250
Fieser 02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
Merck Index 15, 9277
Specific Gravity 0.77
Solubility Information (10% in water) Clear and no haze
Formula Weight 116.21
Percent Purity 99%
Grade Biochemistry
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.