missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine, 98%, MP Biomedicals™

Product Code. 11451612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451612 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451612 Supplier MP Biomedicals Supplier No. 04805613
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 110-18-9
Color Colorless
Density 0.77g/mL at 25°C
Molecular Formula C6H16N2
Quantity 10 mL
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES CN(C)CCN(C)C
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
Molecular Weight (g/mol) 116.208
PubChem CID 8037
ChEBI CHEBI:32850
Formula Weight 116.2
Percent Purity 0.98
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.