missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphat

Product Code. 17251146 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17251146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17251146 Supplier Thermo Scientific Acros Supplier No. 439070250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94790-35-9 | C5H12ClN2 | 135.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 94790-35-9
Melting Point 100.0°C to 106.0°C
Molecular Formula C5H12ClN2
Quantity 25 g
InChI Key QZFWNEASXBNLCD-UHFFFAOYSA-N
SMILES CN(C)C(=[Cl+])N(C)C
IUPAC Name [bis(dimethylamino)methylidene]-λ³-chloranylium
Molecular Weight (g/mol) 135.61
Formula Weight 280.58
Chemical Name or Material N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N,N,N',N'-Tetramethylchloroformamidiniumhexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.