missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine Dihydrochloride, TRC

Product Code. 30455448
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455448 10 g 10g
30454378 25 g 25g
30454823 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455448 Supplier Toronto Research Chemicals Supplier No. TRCT88956810G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H16N2 . 2(HCl)
Molecular Weight (g/mol) 237.17
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES Cl.Cl.CN(C)c1ccc(cc1)N(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H16N2.2ClH/c1-11(2)9-5-7-10(8-6-9)12(3)4;;/h5-8H,1-4H3;2*1H
Formula Weight 236.0847
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.