missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

Product Code. 11440033
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440033 50 g 50g
11430033 10 g 10g
11450033 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440033 Supplier Thermo Scientific Alfa Aesar Supplier No. A12107.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-01-4 | C10H18Cl2N2 | 237.168 g/mol

N,N,N',N'-tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is an oxidizable compound that serves as a reducing co-substrate for heme peroxidases. It is also used as a test reagent in microbiology for the classification of cytochrome oxidase positive aerobic microorganisms. It finds application as a detection of peroxidases on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is an oxidizable compound that serves as a reducing co-substrate for heme peroxidases. It is also used as a test reagent in microbiology for the classification of cytochrome oxidase positive aerobic microorganisms. It finds application as a detection of peroxidases on polyacrylamide gels.

Solubility
Soluble in dimethyl sulfoxide, water and phosphate buffered saline at pH 7.2.

Notes
Light sensitive. Incompatible with strong oxidizing agents.                    
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
MDL Number MFCD00012482
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

Melting Point ∼228°C (decomposition)
Assay Percent Range 99%
Quantity 50 g
Beilstein 5132096
Sensitivity Light sensitive
Solubility Information Soluble in dimethyl sulfoxide,water and phosphate buffered saline at pH 7.2.
Formula Weight 237.17
Percent Purity ≥98%
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: N,N,N',N'-Tetramethyl-p-phenylenediaminedihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.