missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog Number 10327640
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
100-22-1
C10H16N2
164.25
CJAOGUFAAWZWNI-UHFFFAOYSA-N
wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb
7490
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine
CN(C)C1=CC=C(C=C1)N(C)C
This item is not returnable. View return policy
100-22-1
C10H16N2
164.25
CJAOGUFAAWZWNI-UHFFFAOYSA-N
wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb
7490
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine
CN(C)C1=CC=C(C=C1)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48.0°C to 52.0°C
Color Beige-Brown to Brown
Boiling Point 260.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4[N(CH3)2]2
Quantity 5 g
Beilstein 13,74
Merck Index 15,9376
Solubility Information Solubility in water: slightly in cold, more in hot water. Other solubilities: soluble in alcohols, chloroform, ether
Formula Weight 164.25
Percent Purity 98%
Physical Form Crystalline Flakes
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'