missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
Coupling reagent
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 213°C to 215°C |
Color | White |
Elemental Analysis | 29.34 to 30.85% C, 11.41 to 11.99% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Quantity | 100 g |
IUPAC Name | [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate |
Formula Weight | 359.21 |
Show More |
Safety and Handling
Product Identifier
- HSTU
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'