missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Product Code. 15495255
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495255 5 g 5g
15485255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495255 Supplier Thermo Scientific Alfa Aesar Supplier No. H54144.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 125700-69-8 | C12H16BF4N5O2 | 349.10 g/mol

O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N?,N?-tetramethyluronium tetrafluoroborate (2-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, TBTU) may be used as coupling reagent in the synthesis of macrocyclic polyamine derivatives with various length of linkers, condensation reagent in the large-scale preparation of the hematoregulatory nonapeptide (Glp-Glu-Asp)2-DAS-(Lys)2 (2,SK&F 107647), coupling reagent for peptide synthesis which best suppresses racemization during fragment condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate (2-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, TBTU) may be used as coupling reagent in the synthesis of macrocyclic polyamine derivatives with various length of linkers, condensation reagent in the large-scale preparation of the hematoregulatory nonapeptide (Glp-Glu-Asp)2-DAS-(Lys)2 (2,SK&F 107647), coupling reagent for peptide synthesis which best suppresses racemization during fragment condensation.

Solubility
Soluble in acetonitrile 0.1 g/mL.

Notes
Moisture and light sensitive. Store away from water/ moisture, oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125700-69-8
Molecular Formula C12H16BF4N5O2
Molecular Weight (g/mol) 349.10
MDL Number MFCD00077730
InChI Key FOBCPCIJLQTYBT-UHFFFAOYSA-N
Synonym tdbtu, n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-4-oxobenzo d 1,2,3 triazin-3 4h-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, tdbtu; n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, dimethylamino 4-oxo-1,2,3-benzotriazin-3-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12760, acmc-1cac0, d04fkx
PubChem CID 10882602
IUPAC Name [dimethylamino-[(4-oxo-1,2,3-benzotriazin-3-yl)oxy]methylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C

Specifications

Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in acetonitrile 0.1g/mL.
Formula Weight 349.09
Percent Purity ≥98%
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.