missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluo, Thermo Scientific™

Product Code. 11996701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11996701 5 g 5g
11986701 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11996701

Brand: Thermo Scientific Acros 440630050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 125700-69-8
Molecular Formula C12H16BF4N5O2
Molecular Weight (g/mol) 349.10
MDL Number MFCD00077730
InChI Key FOBCPCIJLQTYBT-UHFFFAOYSA-N
Synonym tdbtu, n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-4-oxobenzo d 1,2,3 triazin-3 4h-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, tdbtu; n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, dimethylamino 4-oxo-1,2,3-benzotriazin-3-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12760, acmc-1cac0, d04fkx
PubChem CID 10882602
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 130.0°C
Color White to Yellow
Elemental Analysis 40.67 to 41.91% C, 19.76 to 20.36% N
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 5 g
IUPAC Name [dimethylamino-[(4-oxo-1,2,3-benzotriazin-3-yl)oxy]methylidene]-dimethylazanium;tetrafluoroborate
Formula Weight 349.1
Percent Purity 99%
Physical Form Powder or Solid
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • TDBTU
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.