missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluo, Thermo Scientific™

Product Code. 11986701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11986701 1 g 1g
11996701 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986701 Supplier Thermo Scientific Acros Supplier No. 440630010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 125700-69-8
Molecular Formula C12H16BF4N5O2
Molecular Weight (g/mol) 349.10
MDL Number MFCD00077730
InChI Key FOBCPCIJLQTYBT-UHFFFAOYSA-N
Synonym tdbtu, n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-4-oxobenzo d 1,2,3 triazin-3 4h-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, tdbtu; n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, dimethylamino 4-oxo-1,2,3-benzotriazin-3-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12760, acmc-1cac0, d04fkx
PubChem CID 10882602
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Elemental Analysis 40.67 to 41.91% C, 19.76 to 20.36% N
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 1 g
IUPAC Name [(dimethylamino)[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Formula Weight 349.1
Physical Form Powder or Solid
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • TDBTU
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.