missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-D-tartaramide, TRC

Product Code. 30456074
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456074 1 g 1g
30449532 10 g 10g
30450569 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456074 Supplier Toronto Research Chemicals Supplier No. TRCT3053501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63126-52-3
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.22
Synonym (2S,3S)-2,3-Dihydroxy-N,N,N',N'-tetramethyl-butanediamide, [S-(R*,R*)]-2,3-Dihydroxy-N,N,N',N'-tetramethyl-butanediamide, (-)-D-Tartaric Acid Bis(N,N-dimethylamide), (-)-Tetramethyl-(D)-tartaric Acid Diamide, N,N,N',N'-Tetramethyl-(S,S)-tartaramide
IUPAC Name (2S,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
SMILES CN(C)C(=O)[C@@H](O)[C@H](O)C(=O)N(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H16N2O4/c1-9(2)7(13)5(11)6(12)8(14)10(3)4/h5-6,11-12H,1-4H3/t5-,6-/m0/s1
Formula Weight 204.111
Chemical Name or Material N,N,N',N'-Tetramethyl-D-tartaramide
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.