missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,8-naphthalenediamine, ≥99.0% (GC), Honeywell™ Fluka™

Product Code. 15656100 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Poly bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15656100 1 g Poly bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15656100 Supplier Honeywell-Fluka Supplier No. 641991G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 20734-58-1
Melting Point 49°C to 51°C
Color White
Flash Point 113°C (235.4°F)
Packaging Poly bottle
Molecular Formula C14H18N2
MDL Number MFCD00003920
Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 396782
Synonym 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
InChI Key GJFNRSDCSTVPCJ-UHFFFAOYSA-N
SMILES CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C
IUPAC Name 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
Molecular Weight (g/mol) 214.312
PubChem CID 88675
Formula Weight 214.31g/mol
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine, Matrix substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.