missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,3-propanediamine, 99+%

Product Code. 10647314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10647314 250 g Glass bottle 250g
10133540 10 g Glass bottle 10g
10429450 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10647314 Supplier Thermo Scientific Acros Supplier No. 155512500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-95-2 | C7H18N2 | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-95-2
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.23
MDL Number MFCD00008337
InChI Key DMQSHEKGGUOYJS-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r
PubChem CID 8084
IUPAC Name N,N,N',N'-tetramethylpropane-1,3-diamine
SMILES CN(C)CCCN(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.7800g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)3N(CH3)2
Refractive Index 1.4224 to 1.4244
Quantity 250 g
Beilstein 04, 262
Specific Gravity 0.78
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,3-propanediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-1,3-propanediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.