missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine, Thermo Scientific Chemicals

Product Code. p-7042890
Change view
Click to view available options
Quantity:
100 mg
300 mg
Unit Size:
100mg
300mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470347 100 mg 100mg
15460347 300 mg 300mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470347 Supplier Thermo Scientific Alfa Aesar Supplier No. J64206.MC
Add to basket
This item is not returnable. View return policy

A cell permeable and extremely high-affinity chelator of heavy metals

A cell permeable and extremely high-affinity chelator of heavy metals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16858-02-9
Molecular Formula C26H28N6
Molecular Weight (g/mol) 424.55
MDL Number MFCD00036918
InChI Key CVRXLMUYFMERMJ-UHFFFAOYSA-N
Synonym tpen, n,n,n',n'-tetrakis 2-pyridylmethyl ethylenediamine, n1,n1,n2,n2-tetrakis pyridin-2-ylmethyl ethane-1,2-diamine, unii-r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridylmethyl ethane-1,2-diamine, r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridyl-methyl ethylenediamine, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl-, oc-6-11, 1,2-ethanediamine,n,n,n',n'-tetrakis 2-pyridinylmethyl, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl
PubChem CID 5519
SMILES C(CN(CC1=CC=CC=N1)CC1=CC=CC=N1)N(CC1=CC=CC=N1)CC1=CC=CC=N1

Specifications

Melting Point 110°C to 112°C
Color White
Quantity 100 mg
Beilstein 4211982
IUPAC Name (2-{bis[(pyridin-2-yl)methyl]amino}ethyl)bis[(pyridin-2-yl)methyl]amine
Formula Weight 424.54
Physical Form Powder
Chemical Name or Material N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine
compliance-icons
Product Identifier
  • N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N,N',N'-Tetrakis-(2-pyridylmethyl)ethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.