missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%

Product Code. 10147333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147333 1 kg Glass bottle 1kg
10107383 100 g Glass bottle 100g
10085950 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147333 Supplier Thermo Scientific Acros Supplier No. 420480010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-07-8 | C10H24N2O4 | 236.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-07-8
Molecular Formula C10H24N2O4
Molecular Weight (g/mol) 236.31
MDL Number MFCD00002854
InChI Key BYACHAOCSIPLCM-UHFFFAOYSA-N
Synonym theed, n,n,n',n'-tetrakis 2-hydroxyethyl ethylenediamine, ethylenediamine tetraethanol, entol, tked, ethylenedinitrilotetraethanol, tetrakis hydroxyethyl ethylenediamine, tetra hydroxyethyl ethylenediamine, unii-k5ape098zi, tetrahydroxyethyl ethylenediamine
PubChem CID 67322
SMILES OCCN(CCO)CCN(CCO)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Amber
Density 1.1300g/mL
pH ≥10 (25% soln.)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
Refractive Index 1.4990 to 1.5070
Quantity 1 kg
Specific Gravity 1.13
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
IUPAC Name 2-[2-[bis(2-hydroxyethyl)amino]ethyl-(2-hydroxyethyl)amino]ethanol
Formula Weight 236.31
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.