missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 10107383
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-07-8
Molecular Formula C10H24N2O4
Molecular Weight (g/mol) 236.31
MDL Number MFCD00002854
InChI Key BYACHAOCSIPLCM-UHFFFAOYSA-N
Synonym theed, n,n,n',n'-tetrakis 2-hydroxyethyl ethylenediamine, ethylenediamine tetraethanol, entol, tked, ethylenedinitrilotetraethanol, tetrakis hydroxyethyl ethylenediamine, tetra hydroxyethyl ethylenediamine, unii-k5ape098zi, tetrahydroxyethyl ethylenediamine
PubChem CID 67322
SMILES OCCN(CCO)CCN(CCO)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Amber
Density 1.1300g/mL
pH ≥10 (25% soln.)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
Refractive Index 1.4990 to 1.5070
Quantity 100 g
Specific Gravity 1.13
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
IUPAC Name 2-[2-[bis(2-hydroxyethyl)amino]ethyl-(2-hydroxyethyl)amino]ethanol
Formula Weight 236.31
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'