missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethylethylenediamine, 99+%

Product Code. 10133590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10133590 100 mL 100mL
10307860 10 mL 10mL
11925191 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133590 Supplier Thermo Scientific Acros Supplier No. 155591000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-77-6 | C10H26N2 | 174.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-77-6
Molecular Formula C10H26N2
Molecular Weight (g/mol) 174.33
MDL Number MFCD00009055
InChI Key DIHKMUNUGQVFES-UHFFFAOYSA-P
Synonym n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
PubChem CID 67423
SMILES CC[NH+](CC)CC[NH+](CC)CC

Specifications

Color Yellow
Density 0.8080g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Refractive Index 1.4333 to 1.4353
Quantity 100 mL
Beilstein 04,251
Specific Gravity 0.808
Solubility Information Solubility in water: soluble.
IUPAC Name [2-(diethylazaniumyl)ethyl]diethylazanium
Formula Weight 172.31
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetraethylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.