missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethylethylenediamine, 98%, Thermo Scientific™

Product Code. 10770951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10770951 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10770951 Supplier Thermo Scientific Acros Supplier No. 293420250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-77-6
Molecular Formula C10H26N2
Molecular Weight (g/mol) 174.33
MDL Number MFCD00009055
InChI Key DIHKMUNUGQVFES-UHFFFAOYSA-P
Synonym n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
PubChem CID 67423
IUPAC Name [2-(diethylazaniumyl)ethyl]diethylazanium
SMILES CC[NH+](CC)CC[NH+](CC)CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.8080g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Refractive Index 1.4333 to 1.4353
Quantity 25 mL
Beilstein 04, 251
Specific Gravity 0.808
Solubility Information Solubility in water: soluble.
Formula Weight 172.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetraethylethylenediamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.