missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylenebisacrylamide, 96%, extra pure, Thermo Scientific Chemicals

Product Code. 11985461
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point 185°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Plastic bottle
Linear Formula (H2C=CHCONH)2CH2
Quantity 2.5 kg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 154.17
Percent Purity 96%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material N, N'-Methylenebisacrylamide
Show More Show Less
compliance-icons
Product Identifier
  • N,N'-Methylenebisacrylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'