missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Methylenebis(acrylamide), TRC

Product Code. 30472095
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30472095 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30472095

Brand: Toronto Research Chemicals TRCM30374550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Methylenebis(acrylamide)
CAS 110-26-9
Molecular Formula C7 H10 N2 O2
Quantity 50 g
Synonym Bisacrylamide, MBA, MBAA, Methylenebisacrylamide, Methylenediacrylamide, N,N'-Diacryloylmethylenediamine, N,N'-Methylenebis(2-propenamide), N,N'-Methylenebis(acrylamide), N,N'-Methylenediacrylamide, NMBA, NSC 406836, NSC 7774, Triam 507
InChI Formula InChI=1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)
SMILES C=CC(=O)NCNC(=O)C=C
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
Molecular Weight (g/mol) 154.17
Formula Weight 154.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.