missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Heptyl)-n-octylamine, 98+%, Thermo Scientific™

Product Code. 11389525
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389525 1 g 1g
11399525 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389525 Supplier Thermo Scientific Alfa Aesar Supplier No. L18870.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26627-77-0
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.436
MDL Number MFCD03093076
InChI Key HAEBNPLWJOGFDQ-UHFFFAOYSA-N
Synonym n-n-heptyl-n-octylamine, heptyloctylamine, n-heptyloctylamine, heptyl-octyl-amine, n-heptyl-n-octylamine, heptyl octyl amine, 1-octanamine, n-heptyl
PubChem CID 12905714
IUPAC Name N-heptyloctan-1-amine
SMILES CCCCCCCCNCCCCCCC

Specifications

Refractive Index 1.442
Quantity 1 g
UN Number UN2735
Beilstein 2346655
Sensitivity Air Sensitive
Formula Weight 227.44
Percent Purity ≥98%
Chemical Name or Material N-(n-Heptyl)-n-octylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.