missing translation for 'onlineSavingsMsg'
Learn More

N-(N-Heptyl)-5-chloro-1-naphthalenesulfonamide, TRC

Product Code. 30397490
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397490 10 mg 10mg
30388786 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397490 Supplier Toronto Research Chemicals Supplier No. TRCH28300010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102649-79-6
Molecular Formula C17 H22 Cl N O2 S
Molecular Weight (g/mol) 339.88
IUPAC Name 5-chloro-N-heptylnaphthalene-1-sulfonamide
SMILES CCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H22ClNO2S/c1-2-3-4-5-6-13-19-22(20,21)17-12-8-9-14-15(17)10-7-11-16(14)18/h7-12,19H,2-6,13H2,1H3
Formula Weight 339.106
Chemical Name or Material N-(N-Heptyl)-5-chloro-1-naphthalenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.