missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Ethylenebisacrylamide, 96%

Product Code. 11383226
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11383226

Brand: Thermo Scientific Alfa Aesar L12544.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2956-58-3 | C8H12N2O2 | 168.196 g/mol

N,N'-Ethylenebisacrylamide is used in the preparation of N,N'-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Ethylenebisacrylamide is used in the preparation of N,N′-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2956-58-3
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00042935
InChI Key AYGYHGXUJBFUJU-UHFFFAOYSA-N
Synonym n,n'-ethylenebisacrylamide, n,n-ethylenebisacrylamide, ethylene bis acrylamide, n,n'-ethane-1,2-diyl diacrylamide, 2-propenamide, n,n'-1,2-ethanediylbis, n,n'-bis acryloyl-1,2-diaminoethane, n,n'-bis acryloyl-1,2-diaminoethane, stabilized with hydroquinone, n-2-prop-2-enoylamino ethyl prop-2-enamide, n-2-prop-2-enamido ethyl prop-2-enamide, ethylenebisacrylamide
PubChem CID 168964
IUPAC Name N-[2-(prop-2-enoylamino)ethyl]prop-2-enamide
SMILES C=CC(=O)NCCNC(=O)C=C

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Beilstein 1771570
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
Formula Weight 168.2
Percent Purity 96%
Chemical Name or Material N,N'-Ethylenebisacrylamide
compliance-icons
Product Identifier
  • N,N'-Ethylenebisacrylamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Ethylenebis(acrylamide)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.