missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
N,N'-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.
Solubility
Soluble in dimethyl sulfoxide, hot pyridine, acetonitrile and most organic solvents. Insoluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids and oxidizing agents.
N,N′-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.
Solubility
Soluble in dimethyl sulfoxide, hot pyridine, acetonitrile and most organic solvents. Insoluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids and oxidizing agents.
Specifications
Melting Point | 190°C (decomposition) |
Quantity | 1 g |
Beilstein | 1499137 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in dimethyl sulfoxide,hot pyridine,acetonitrile and most organic solvents. Insoluble in water. |
Formula Weight | 256.17 |
Percent Purity | 85% |
Grade | Technical |
Assay | remainder N-Hydroxysuccinimide |
Chemical Name or Material | N,N'-Disuccinimidyl carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'