missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Dipropylurea, TRC

Product Code. 30381624
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381624 25 g
30384494 1 g
30380548 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381624 Supplier Toronto Research Chemicals Supplier No. TRCD49246025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-95-0
Molecular Formula C7 H16 N2 O
Molecular Weight (g/mol) 144.215
Synonym N,N'-Dipropylurea, Urea, 1,3-dipropyl- (6CI,7CI,8CI), 1,3-Di-n-propylurea, 1,3-Dipropylurea, N,N'-Dipropylurea, Chlorpropamide Imp. B (EP 7.0)
IUPAC Name 1,3-dipropylurea
SMILES CCCNC(=O)NCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H16N2O/c1-3-5-8-7(10)9-6-4-2/h3-6H2,1-2H3,(H2,8,9,10)
Formula Weight 144.126
Chemical Name or Material 1,3-Dipropylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.