missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylformamidine, TRC

Product Code. 30385373
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385373 5 g
30382261 50 g
30382840 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385373 Supplier Toronto Research Chemicals Supplier No. TRCD4910005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-15-1
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.25
Synonym N,N'-Diphenylmethanimidamide, 1,3-Diphenyl-1,3-diaza-1-propene, Diphenylformamidine
SMILES N(\C=N\c1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H12N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h1-11H,(H,14,15)
Chemical Name or Material N,N'-Diphenylformamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.