missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diphenylcarbamyl Chloride, TRC

Product Code. 30383545
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383545 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383545 Supplier Toronto Research Chemicals Supplier No. TRCD48734010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylcarbamic Chloride
CAS 83-01-2
Molecular Formula C13 H10 Cl N O
Quantity 10 g
Synonym Diphenylcarbamic Chloride, Temozolomide Related Compound C (USP)
InChI Formula InChI=1S/C13H10ClNO/c14-13(16)15(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
SMILES ClC(=O)N(c1ccccc1)c2ccccc2
IUPAC Name N,N-diphenylcarbamoyl chloride
Molecular Weight (g/mol) 231.68
Formula Weight 231.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.