missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylbenzidine, 98%

Product Code. 11494874
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494874 5 g 5g
11404884 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494874 Supplier Thermo Scientific Alfa Aesar Supplier No. A13802.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 531-91-9 | C24H20N2 | 336.44 g/mol

N,N'-Diphenylbenzidine is an indicator suitable for redox titrations, e.g. with dichromate or cerium(IV) sulfate 1,2. It is also used as intermediates in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diphenylbenzidine is an indicator suitable for redox titrations, e.g. with dichromate or cerium(IV) sulfate 1,2. It is also used as intermediates in organic synthesis.

Solubility
Insoluble in water.

Notes
Light Sensitive. Do not store above 23°C. Store away from Strong bases. Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-91-9
Molecular Formula C24H20N2
Molecular Weight (g/mol) 336.44
MDL Number MFCD00003016
InChI Key FDRNXKXKFNHNCA-UHFFFAOYSA-N
Synonym n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
PubChem CID 68280
IUPAC Name 4-(4-anilinophenyl)-N-phenylaniline
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 246°C to 250°C
Odor Odorless
Quantity 5 g
Beilstein 2220993
Sensitivity Light sensitive
Merck Index 14,3319
Solubility Information Insoluble in water.
Formula Weight 336.44
Percent Purity 98%
Chemical Name or Material N,N'-Diphenylbenzidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.