missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-p-phenylenediamine, 97%

Product Code. 11460138
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460138 25 g 25g
11450138 5 g 5g
11470138 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460138 Supplier Thermo Scientific Alfa Aesar Supplier No. B23713.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 74-31-7 | C18H16N2 | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-31-7
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00003015
InChI Key UTGQNNCQYDRXCH-UHFFFAOYSA-N
Synonym n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
PubChem CID 6319
ChEBI CHEBI:34860
IUPAC Name 1-N,4-N-diphenylbenzene-1,4-diamine
SMILES N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 146°C to 153°C
Density 1.28
Boiling Point 220°C to 225°C (1 mmHg)
Quantity 25 g
Merck Index 14,3329
Formula Weight 260.34
Percent Purity 97%
Chemical Name or Material N,N'-Diphenyl-p-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.