missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyltrimethylsilylamine, 97%

Product Code. 10751535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751535 50 mL Glass bottle 50mL
10328730 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751535 Supplier Thermo Scientific Acros Supplier No. 209970500
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2083-91-2 | C5H15NSi | 117.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2083-91-2
Molecular Formula C5H15NSi
Molecular Weight (g/mol) 117.27
MDL Number MFCD00008297
InChI Key KAHVZNKZQFSBFW-UHFFFAOYSA-N
Synonym n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
PubChem CID 74965
IUPAC Name N-methyl-N-trimethylsilylmethanamine
SMILES CN(C)[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7320g/mL
Boiling Point 84°C
Flash Point -19°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)3SiN(CH3)2
Refractive Index 1.3960 to 1.398
Quantity 50 mL
Specific Gravity 0.732
Solubility Information Solubility in water: decomposes
Formula Weight 117.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyltrimethylsilylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyltrimethylsilylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.