missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylthiourea, 99%, Thermo Scientific Chemicals

Product Code. 10386550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10386550 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386550 Supplier Thermo Scientific Acros Supplier No. 116890050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 534-13-4
Molecular Formula C3H8N2S
Molecular Weight (g/mol) 104.18
MDL Number MFCD00004923
InChI Key VLCDUOXHFNUCKK-UHFFFAOYSA-N
Synonym n,n'-dimethylthiourea, 1,3-dimethyl-2-thiourea, thiourea, n,n'-dimethyl, sym-dimethylthiourea, dimethylthiocarbamide, dimethylthiourea, 1,3-dimethylisothiourea, dimethyl-2-thiourea, urea, 1,3-dimethyl-2-thio, unii-8p30pmd17w
PubChem CID 2723631
IUPAC Name 1,3-dimethylthiourea
SMILES CNC(=S)NC

Specifikationer

CAS Min % 98.5
CAS Max % 100.0
Melting Point 61°C to 67°C
Color Colorless to White
Boiling Point 155°C (1.0 Mbar)
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3NHCSNHCH3
Quantity 5 g
Beilstein 04, 70
Merck Index 15, 3287
Solubility Information Solubility in water: 2900g/L (20°C). Other solubilities: soluble in alcohol and acetone,sparingly soluble in benzene,ether and carbon,disulfide,slightly soluble in petroleum ether
Formula Weight 104.18
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N, N'-Dimethylthiourea, 99%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • N,N'-Dimethylthiourea
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Faroangivelser
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.