missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylthiourea, 99%, Thermo Scientific Chemicals

Product Code. p-120750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10386550

Brand: Thermo Scientific Acros 116890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-13-4
Molecular Formula C3H8N2S
Molecular Weight (g/mol) 104.18
MDL Number MFCD00004923
InChI Key VLCDUOXHFNUCKK-UHFFFAOYSA-N
Synonym n,n'-dimethylthiourea, 1,3-dimethyl-2-thiourea, thiourea, n,n'-dimethyl, sym-dimethylthiourea, dimethylthiocarbamide, dimethylthiourea, 1,3-dimethylisothiourea, dimethyl-2-thiourea, urea, 1,3-dimethyl-2-thio, unii-8p30pmd17w
PubChem CID 2723631
IUPAC Name 1,3-dimethylthiourea
SMILES CNC(=S)NC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 61°C to 67°C
Color Colorless to White
Boiling Point 155°C (1.0 Mbar)
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3NHCSNHCH3
Quantity 5 g
Beilstein 04, 70
Merck Index 15, 3287
Solubility Information Solubility in water: 2900g/L (20°C). Other solubilities: soluble in alcohol and acetone,sparingly soluble in benzene,ether and carbon,disulfide,slightly soluble in petroleum ether
Formula Weight 104.18
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N, N'-Dimethylthiourea, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N,N'-Dimethylthiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.