missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyltetradecylamine, TRC

Product Code. 30380901
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380901 50 g -
30379809 25 g 25g
30386023 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380901 Supplier Toronto Research Chemicals Supplier No. TRCD47948050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-75-4
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
Synonym N,N-Dimethyl-1-tetradecanamine, Dimethylmyristamine, Farmin DM 4098, Genamin 14R302D, IPL 30, Myristyldimethylamine, N-Tetradecyldimethylamine, NSC 78319, Tetradecyldimethylamine
IUPAC Name N,N-dimethyltetradecan-1-amine
SMILES CCCCCCCCCCCCCCN(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C16H35N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17(2)3/h4-16H2,1-3H3
Formula Weight 241.277
Chemical Name or Material N,N-Dimethyltetradecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.