missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylpropanolamine, TRC

Product Code. 30370161
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370161 100 mL
30371102 500 mL
30376043 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370161 Supplier Toronto Research Chemicals Supplier No. TRCD477830100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-63-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
Synonym 1-Dimethylamino-3-propanol, 3-(Dimethylamino)propanol, 3-(Dimethylamino)propyl Alcohol, 3-(N,N-Dimethylamino)-1-propanol, 3-(N,N-Dimethylamino)propanol, Dimethylpropanolamine, N,N-Dimethyl-3-amino-1-propanol, N,N-Dimethyl-3-aminopropanol, N,N-Dimethyl-3-hydroxypropylamine, N,N-Dimethyl-gamma-aminopropanol, NSC 62086, gamma-(Dimethylamino)propanol
SMILES CN(C)CCCO

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C5H13NO/c1-6(2)4-3-5-7/h7H,3-5H2,1-2H3
Formula Weight 103.0997
Chemical Name or Material N,N-Dimethylpropanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.