missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylpiperazine, TRC

Product Code. 30378142
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378142 1 g
30375908 500 mg
30372818 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378142 Supplier Toronto Research Chemicals Supplier No. TRCD4812981G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6 H14 N2
Molecular Weight (g/mol) 114.19
Synonym N,N'-Dimethylpiperazine, Piperazine, 1,4-dimethyl-, 1,4-Dimethylpiperazine, Addocat 117, Jeffcat DMP, Lupetazine, N,N'-Dimethylpiperazine, NSC 41177, RCA 117, Texacat DMP, AH 1243X
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(C)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H14N2/c1-7-3-5-8(2)6-4-7/h3-6H2,1-2H3
Formula Weight 114.1157
Chemical Name or Material N,N'-Dimethylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.