missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylpiperazine, 98.5%

Product Code. 10783651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10783651 100 mL Glass bottle 100mL
10254380 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10783651 Supplier Thermo Scientific Acros Supplier No. 116651000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-58-1 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00006156
InChI Key RXYPXQSKLGGKOL-UHFFFAOYSA-N
Synonym n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
PubChem CID 7818
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(CC1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -1°C
Color Colorless to Yellow
Density 0.8520g/mL
Boiling Point 130°C to 133°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4453 to 1.4473
Quantity 100 mL
Beilstein 23, 7
Specific Gravity 0.852
Solubility Information Solubility in water: miscible
Viscosity 1.5 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98.50%
Physical Form Liquid
Chemical Name or Material N, N'-Dimethylpiperazine, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N'-Dimethylpiperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.