missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctylamine, 97%

Product Code. 10105163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105163 2.5 L Glass bottle 2.5 L
10206570 25 mL Glass bottle 25mL
10657552 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105163 Supplier Thermo Scientific Acros Supplier No. 225290025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7378-99-6 | C10H23N | 157.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name N,N-dimethyloctan-1-amine
SMILES CCCCCCCCN(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -57.0°C
Density 0.7650g/mL
Boiling Point 195.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (C8-chains) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7N(CH3)2
Refractive Index 1.4233 to 1.4253
Quantity 2.5 L
Beilstein 04,I,386
Specific Gravity 0.765
Solubility Information Solubility in water: slightly soluble
Formula Weight 157.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyloctylamine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.