missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctadecylamine Oxide, TRC

Product Code. 30380889
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380889 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380889 Supplier Toronto Research Chemicals Supplier No. TRCD476200250MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylstearylamine N-Oxide
CAS 2571-88-2
Molecular Formula C20 H43 N O
Quantity 250 mg
Synonym Octadecylamine, N,N-dimethyl-, N-oxide (7CI,8CI), Admox 18, Admox SC 1885, Ammonyx SO, Aromox DM 18D-M, Barlox 18, Barlox 18S, Conco XA 2S, DONO, Dimethyloctadecylamine oxide, Dimethylstearylamine oxide, N,N-Dimethyl-1-octadecanamine oxide, N,N-Dimethyl-N
InChI Formula InChI=1S/C20H43NO/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(2,3)22/h4-20H2,1-3H3
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)[O-]
IUPAC Name N,N-dimethyloctadecan-1-amine oxide
Molecular Weight (g/mol) 313.562
Formula Weight 313.334
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.