missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctadecan-1-amine, TRC

Product Code. 30380251
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380251 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380251 Supplier Toronto Research Chemicals Supplier No. TRCD4770952.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyloctadecan-1-amine
CAS 124-28-7
Molecular Formula C20 H43 N
Quantity 2.5 g
Synonym 1-Octadecanamine, N,N-dimethyl-, Octadecylamine, N,N-dimethyl- (6CI,8CI), N,N-Dimethyl-1-octadecanamine, ADMA 18, AT 1895A, Adogen 342D, Armeen DM 18D, DM 18D, Dimantine, Dimethyl-n-octadecylamine, Dimethyloctadecylamine, Dimethylstearylamine, Dymanthine, Farmin DM 80, Farmin DM 8098, Genamin 18R302D, Kemamine 9902D, N,N-Dimethyl-n-octadecylamine, N,N-Dimethyloctadecan-1-amine, N,N-Dimethyloctadecanamine, N,N-Dimethyloctadecylamine, N,N-Dimethylstearylamine, N-Octadecyl-N,N-dimethylamine, N-Octadecyldimethylamine, N-Stearyl-N,N-dimethylamine, Nissan Tertiary Amine AB, Octadecyldimethylamine, Stearyldimethylamine, Tertiary Nissan Amine AB
InChI Formula InChI=1S/C20H43N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(2)3/h4-20H2,1-3H3
SMILES CCCCCCCCCCCCCCCCCCN(C)C
IUPAC Name N,N-dimethyloctadecan-1-amine
Molecular Weight (g/mol) 297.5621
Formula Weight 297.3396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.