missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexylamine, 99%

Product Code. 11413607
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413607 5 g 5g
11423607 25 g 25g
11433607 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413607 Supplier Thermo Scientific Alfa Aesar Supplier No. A17364.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4385-04-0 | C8H19N | 129.247 g/mol

N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

Solubility
Miscible with methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4385-04-0
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009522
InChI Key QMHNQZGXPNCMCO-UHFFFAOYSA-N
Synonym n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
PubChem CID 78090
IUPAC Name N,N-dimethylhexan-1-amine
SMILES CCCCCCN(C)C

Specifications

Density 0.744
Boiling Point 146°C to 150°C
Flash Point 33°C (92°F)
Refractive Index 1.4136
Quantity 5 g
UN Number UN2733
Solubility Information Miscible with methanol.
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material N,N-Dimethylhexylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.