missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexylamine, 99%

Product Code. 10026403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10026403 25 g Glass bottle 25g
10636111 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10026403 Supplier Thermo Scientific Acros Supplier No. 314740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4385-04-0 | C8H19N | 129.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4385-04-0
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.25
MDL Number MFCD00009522
InChI Key QMHNQZGXPNCMCO-UHFFFAOYSA-N
Synonym n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
PubChem CID 78090
IUPAC Name N,N-dimethylhexan-1-amine
SMILES CCCCCCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7440g/mL
Boiling Point 146°C to 150°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5N(CH3)2
Refractive Index 1.4126 to 1.4146
Quantity 25 g
Specific Gravity 0.744
Formula Weight 129.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylhexylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.