missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylguanidine sulfate, 98%, Thermo Scientific Chemicals

Product Code. 10657941
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-65-2
Molecular Formula 0·5 H2SO4
Molecular Weight (g/mol) 272.32
MDL Number MFCD00013131
InChI Key QSCHFHVDZCPIKX-UHFFFAOYSA-N
Synonym 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1
PubChem CID 69024
IUPAC Name 1,1-dimethylguanidine;sulfuric acid
SMILES CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 280.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH)2NC(NH)NH2)2·H2SO4
Quantity 50 g
Beilstein 04,II,574
Solubility Information Solubility in water: 490g/L (20°C)
Formula Weight 272.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N, N-Dimethylguanidine sulfate, 93%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'