missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylguanidine sulfate, 98%

Product Code. 10442971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442971 100 g Plastic bottle 100g
10730565 10 g Plastic bottle 10g
10657941 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10442971 Supplier Thermo Scientific Acros Supplier No. 408351000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-65-2 | 0·5 H2SO4 | 272.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-65-2
Molecular Formula 0·5 H2SO4
Molecular Weight (g/mol) 272.32
MDL Number MFCD00013131
InChI Key QSCHFHVDZCPIKX-UHFFFAOYSA-N
Synonym 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1
PubChem CID 69024
IUPAC Name 1,1-dimethylguanidine;sulfuric acid
SMILES CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 280.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH)2NC(NH)NH2)2·H2SO4
Quantity 100 g
Beilstein 04,II,574
Solubility Information Solubility in water: 490g/L (20°C)
Formula Weight 272.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N, N-Dimethylguanidine sulfate, 93%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.