missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine, 98+%

Product Code. 11474207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474207 25 g 25g
11464207 5 g 5g
11484207 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474207 Supplier Thermo Scientific Alfa Aesar Supplier No. A19670.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1118-68-9 | C4H9NO2 | 103.121 g/mol

N,N-Dimethylglycine is used as an athletic performance enhancer and immunostimulant. It is also used in the treatment of autism, epilepsy and mitochondrial disease. It is also employed as a biomarker of protein degradation in chronic obstructive lung disease (COPD) patients. Further, it is used as a substrate to identify, differentiate and characterize amino acid methyltransferase. It plays an important role for the development of glycine-based ionic liquids and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-68-9
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00004283
InChI Key FFDGPVCHZBVARC-UHFFFAOYSA-N
Synonym n,n-dimethylglycine, dimethylglycine, 2-dimethylamino acetic acid, dimethylamino acetic acid, n,n-dimethylaminoacetic acid, glycine, n,n-dimethyl, n-methylsarcosine, n,n-dimethyl glycine, unii-7797m4cppa, dmg
PubChem CID 673
ChEBI CHEBI:17724
IUPAC Name 2-(dimethylamino)acetic acid
SMILES CN(C)CC(=O)O

Specifications

Melting Point 179°C to 183°C
pH 6 to 7
Linear Formula (CH3)2NCH2CO2H
Merck Index 14,3244
Solubility Information Soluble in water.
Quantity 25 g
Odor Odorless
Formula Weight 103.12
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylglycine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.