missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals

Catalog Number 11371349
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is suitable for use in ultraviolet spectrophotometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.944g/cm3
Ignition Point 440°C (824°F)
Assay Percent Range >99.7%
Odor Amine-like
Quantity 1 L
Formula Weight 73.09
Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
Grade Sprectrophotometric
Physical Form Liquid
Chemical Name or Material N, N-Dimethylformamide, Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'