missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, purum, Honeywell™

Product Code. 15675390 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15675390 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15675390 Supplier Honeywell Research Chemicals Supplier No. 402552.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 68-12-2
Density 0.944g/mL
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Molecular Formula C3H7NO
Linear Formula HCON(CH3)2
MDL Number MFCD00003284
Vapor Pressure 2.7 mmHg (20°C)
UN Number UN2265
Beilstein 605365
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.10
PubChem CID 6228
ChEBI CHEBI:17741
Formula Weight 73.09g/mol
Percent Purity ≥98% (GC)
Grade purum
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.