missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, HPLC Grade, 99.7+%

Product Code. 11328177
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
6 x 1 L
Unit Size:
1 L
16 L
4 L
6 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328177 4 L 4 L
11318177 1 L 1 L
11338177 4 x 1 L 4 L
11308177 6 x 1 L 6 L
11440510 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11328177 Supplier Thermo Scientific Alfa Aesar Supplier No. 022915.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Color Colorless
Appearance Colorless liquid
Density 0.944 g/cm3 at 20°C
Quantity 4 L
Flash Point 57°C (135°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.431
Assay Percent Range >99.7%
Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
UN Number 2265
Beilstein 605365
Boiling Point 153°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Grade HPLC
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.