missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%

Product Code. 11380034
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380034 5 g 5g
11390034 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380034 Supplier Thermo Scientific Alfa Aesar Supplier No. L00431.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 36805-97-7 | C11H25NO2 | 203.326 g/mol

N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

Solubility
Miscible with toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36805-97-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.326
MDL Number MFCD00015002
InChI Key DBNQIOANXZVWIP-UHFFFAOYSA-N
Synonym n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine
PubChem CID 547712
IUPAC Name N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine
SMILES CC(C)(C)OC(N(C)C)OC(C)(C)C

Specifications

Density 0.85
Boiling Point 75°C to 77°C (12 mmHg)
Flash Point 33°C (91°F)
Linear Formula (CH3)2NCH[OC(CH3)3]2
Refractive Index 1.412
Quantity 5 g
UN Number UN1993
Beilstein 969629
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene and benzene.
Formula Weight 203.33
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Dimethylformamide di-tert-butyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.